Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_145970.1 | NP_666082.2 | 21 | 2167 | |
GenBank mRNA | BC016188.1 | - | 21 | 2167 | |
BC027028.1 | - | 20 | 1982 | 5363547 | |
BC030439.1 | - | 5 | 274 | ||
DQ329239.1 | - | 14 | 1217 |