Gene Transcripts

Gene Symbol
Tdpoz1
Entrez Gene ID
207213
Gene Name
TD and POZ domain containing 1
Gene Aliases
2cpoz56, Mapp2, Spopl1
Chromosome Location
Chr. 3: 93669333 - 93676283 on Build GRCm38
UniGene ID
Mm.425165
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_148949.2 NP_683751.2 2 1391
GenBank mRNA AF290198.1 - 2 1376

View Full Product Details