Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_152922.3 | NP_690886.3 | 2, 3 | 177 | |
GenBank mRNA | AK148723.1 | - | 2, 3 | 178 | |
AY635583.1 | - | 2, 3 | 178 | ||
BC139484.1 | - | 2, 3 | 146 | 8862506 | |
BC141312.1 | - | 2, 3 | 146 | 9055924 | |
BC145632.1 | - | 2, 3 | 146 | 9054200 |