Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_153126.2 | NP_694766.1 | 2 | 293 | |
GenBank mRNA | AK036294.1 | - | 2 | 241 | |
AK040070.1 | - | 2 | 248 | ||
AK054029.1 | - | 2 | 314 | ||
AK087445.1 | - | 2 | 251 | ||
AK142682.1 | - | 2 | 305 | ||
AK149825.1 | - | 2 | 120 | ||
BC034516.1 | - | 2 | 293 | 4458024 |