Gene Transcripts

Gene Symbol
Rtp3
Entrez Gene ID
235636
Gene Name
receptor transporter protein 3
Gene Aliases
Tmem7
Chromosome Location
Chr. 9: 110984938 - 110990578 on Build GRCm38
UniGene ID
Mm.441138
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_153100.2 NP_694740.2 3 2412
GenBank mRNA AJ428064.1 - 3 2403
BC112903.1 - 3 2412 4196964

View Full Product Details