Gene Transcripts

Gene Symbol
Tcstv3
Entrez Gene ID
236219
Gene Name
2-cell-stage, variable group, member 3
Gene Aliases
Chromosome Location
Chr. 13: 120316862 - 120318261 on Build GRCm38
UniGene ID
Mm.302175
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_153523.3 NP_705743.1 2 629
GenBank mRNA AF067060.1 - 627
BC125501.1 - 2 612 40130016
BC125503.1 - 2 612 40130018

View Full Product Details