Gene Transcripts
- Gene Symbol
- VCAM1
- Entrez Gene ID
- 7412
- Gene Name
- vascular cell adhesion molecule 1
- Gene Aliases
- CD106, INCAM-100
- Chromosome Location
- Chr. 1: 100719640 - 100739045 on Build GRCh38
- UniGene ID
- Hs.109225
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001078.3 |
NP_001069.1
|
2, 3 |
556 |
|
NM_080682.2 |
NP_542413.1
|
2, 3 |
556 |
|
GenBank mRNA |
AK223266.1 |
- |
2, 3 |
455 |
|
AK291732.1 |
- |
2, 3 |
454 |
|
AU121762.1 |
- |
2, 3 |
456 |
|
BC017276.2 |
- |
2, 3 |
436 |
4845424 |
BC068490.1 |
- |
2, 3 |
454 |
30339170 |
BC085003.1 |
- |
2, 3 |
436 |
6375943 |
M30257.1 |
- |
2, 3 |
441 |
|
M60335.1 |
- |
2, 3 |
434 |
|
X53051.1 |
- |
2, 3 |
335 |
|
View Full Product Details