Gene Transcripts

Gene Symbol
Tmc7
Entrez Gene ID
209760
Gene Name
transmembrane channel-like gene family 7
Gene Aliases
1700030H01Rik, C230064B05, C630024K23Rik
Chromosome Location
Chr. 7: 118535841 - 118584790 on Build GRCm38
UniGene ID
Mm.69380
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_172476.4 NP_766064.2 16 3842
GenBank mRNA AK083189.1 - 16 3845
BC105966.1 - 10 2856

View Full Product Details