Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_173047.3 | NP_766635.1 | 2 | 438 | |
GenBank mRNA | AK003232.1 | - | 2 | 443 | |
AK028150.1 | - | 2 | 414 | ||
BC028763.1 | - | 2 | 448 | 1313642 | |
BC087735.1 | - | 2 | 399 | 5369996 | |
BC096658.1 | - | 2 | 408 | 5145435 |