Gene Transcripts

Gene Symbol
Vat1l
Entrez Gene ID
270097
Gene Name
vesicle amine transport protein 1 homolog-like (T. californica)
Gene Aliases
9430073I07, AI427515, mKIAA1576
Chromosome Location
Chr. 8: 114205582 - 114374074 on Build GRCm38
UniGene ID
Mm.334825
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_173016.3 NP_766604.2 9 2019
GenBank mRNA AK035009.1 - 2 2610
AK122527.1 - 9 2019
BC056927.1 - 9 2029 5719211

View Full Product Details