Gene Transcripts

Gene Symbol
Wdr27
Entrez Gene ID
71682
Gene Name
WD repeat domain 27
Gene Aliases
0610012K18Rik
Chromosome Location
Chr. 17: 14784832 - 14943151 on Build GRCm38
UniGene ID
Mm.122698
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_175173.3 NP_780382.2 3 245
GenBank mRNA AK077050.1 - 3 246

View Full Product Details