Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_175325.3 | NP_780534.1 | 3, 4 | 239 | |
GenBank mRNA | AK039560.1 | - | 3, 4 | 188 | |
AK089970.1 | - | 3, 4 | 194 | ||
AK134734.1 | - | 3, 4 | 192 | ||
BC055797.1 | - | 3, 4 | 159 | ||
BC089507.1 | - | 3, 4 | 173 | ||
BC092531.1 | - | 3, 4 | 183 | ||
BC145771.1 | - | 3, 4 | 201 | 40131074 | |
BC145773.1 | - | 3, 4 | 201 | 40131076 |