Gene Transcripts

Gene Symbol
Lyrm2
Entrez Gene ID
108755
Gene Name
LYR motif containing 2
Gene Aliases
2610208E05Rik
Chromosome Location
Chr. 4: 32800259 - 32802268 on Build GRCm38
UniGene ID
Mm.1182
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_175364.4 NP_780573.1 3 969
GenBank mRNA AK044791.1 - 3 965

View Full Product Details