Gene Transcripts

Gene Symbol
Rab11fip4
Entrez Gene ID
268451
Gene Name
RAB11 family interacting protein 4 (class II)
Gene Aliases
A730072L08Rik, RAB11-FIP4, mKIAA1821
Chromosome Location
Chr. 11: 79591021 - 79698023 on Build GRCm38
UniGene ID
Mm.130656
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_175543.3 NP_780752.1 1, 2 223
GenBank mRNA AK046734.1 - 1, 2 225
AK147219.1 - 1, 2 294
AK147344.1 - 1, 2 420

View Full Product Details