Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001195265.1 | NP_001182194.1 | 16 | 2939 | |
GenBank mRNA | AK033653.1 | - | 6 | 834 | |
BC096488.1 | - | 6 | 1055 | ||
BC147398.1 | - | 3 | 742 | 9056452 | |
BC147399.1 | - | 3 | 742 | 9056443 | |
BC172054.1 | - | 3 | 742 | ||
KF041446.1 | - | 17 | 2905 | ||
KF041447.1 | - | 17 | 2934 | ||
KF041448.1 | - | 14 | 2719 | ||
KF041449.1 | - | 15 | 2752 | ||
KF041450.1 | - | 16 | 2844 | ||
KF041451.1 | - | 17 | 2725 |