Gene Transcripts

Gene Symbol
Ralyl
Entrez Gene ID
76897
Gene Name
RALY RNA binding protein-like
Gene Aliases
0710005M24Rik
Chromosome Location
Chr. 3: 13471655 - 14182287 on Build GRCm38
UniGene ID
Mm.422625
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001163328.1 NP_001156800.1 2 385
NM_178631.4 NP_848746.1 3 478
GenBank mRNA AK032086.1 - 2 318
AK079659.1 - 2 1207

View Full Product Details