Gene Transcripts

Gene Symbol
ACVR2B
Entrez Gene ID
93
Gene Name
activin A receptor type 2B
Gene Aliases
ACTRIIB, ActR-IIB, HTX4
Chromosome Location
Chr. 3: 38454299 - 38493142 on Build GRCh38
UniGene ID
Hs.174273
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001106.3 NP_001097.2 9 1176
XM_005265583.3 XP_005265640.1 9 3638
XM_017007514.1 XP_016863003.1 9 3617
XM_017007515.1 XP_016863004.1 9 1262
XM_017007516.1 XP_016863005.1 9 4586
GenBank mRNA AB209449.1 - 8 4941
BC096243.1 - 9 1156 40005760
BC096244.1 - 9 1156 40005761
BC096245.1 - 8 1238 40005763
BC099642.4 - 9 1156 40005762
X77533.1 - 9 1156

View Full Product Details