Gene Transcripts
- Gene Symbol
- PTTG1IP
- Entrez Gene ID
- 754
- Gene Name
- pituitary tumor-transforming 1 interacting protein
- Gene Aliases
- C21orf1, C21orf3, PBF
- Chromosome Location
- Chr. 21: 44849585 - 44873903 on Build GRCh38
- UniGene ID
- Hs.474010
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_004339.3 |
NP_004330.1
|
3, 4 |
572 |
|
GenBank mRNA |
AF149785.1 |
- |
3, 4 |
510 |
|
AK094882.1 |
- |
3, 4 |
305 |
|
AK290139.1 |
- |
3, 4 |
325 |
|
AK315626.1 |
- |
3, 4 |
387 |
|
BC000415.2 |
- |
3, 4 |
329 |
2961420 |
BC012858.1 |
- |
3, 4 |
355 |
3857505 |
BC019295.1 |
- |
3, 4 |
319 |
2823814 |
BC020983.1 |
- |
3, 4 |
336 |
3851447 |
BC031097.1 |
- |
3, 4 |
350 |
5288388 |
BC034250.1 |
- |
3, 4 |
344 |
3862830 |
BG721942.1 |
- |
3, 4 |
511 |
|
Z50022.1 |
- |
3, 4 |
378 |
|
View Full Product Details