Gene Transcripts

Gene Symbol
CASP3
Entrez Gene ID
836
Gene Name
caspase 3
Gene Aliases
CPP32, CPP32B, SCA-1
Chromosome Location
Chr. 4: 184627696 - 184649475 on Build GRCh38
UniGene ID
Hs.141125
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_004346.3 NP_004337.2 4 361
NM_032991.2 NP_116786.1 3 194
XM_011532301.1 XP_011530603.1 4 295
GenBank mRNA AK291337.1 - 4 332
AU127028.1 - 3 194
AU133051.1 - 4 333
BC016926.2 - 4 313 4419175
BX647609.1 - 4 433
U13737.1 - 4 322
U13738.1 - 3 161
U26943.1 - 2 98

View Full Product Details