Gene Transcripts
- Gene Symbol
- TIMM8A
- Entrez Gene ID
- 1678
- Gene Name
- translocase of inner mitochondrial membrane 8 homolog A (yeast)
- Gene Aliases
- DDP, DDP1, DFN1, MTS, TIM8
- Chromosome Location
- Chr. X: 101345656 - 101348969 on Build GRCh38
- UniGene ID
- Hs.447877
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
U66035.1 |
- |
1 |
86 |
|
View Full Product Details