Gene Transcripts

Gene Symbol
DMP1
Entrez Gene ID
1758
Gene Name
dentin matrix acidic phosphoprotein 1
Gene Aliases
ARHP, ARHR, DMP-1
Chromosome Location
Chr. 4: 87650302 - 87664361 on Build GRCh38
UniGene ID
Hs.652366
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001079911.2 NP_001073380.1 2 116
NM_004407.3 NP_004398.1 2 116
XM_011531705.2 XP_011530007.1 2 991
XM_011531706.2 XP_011530008.1 2 991
GenBank mRNA BC130581.1 - 1 38
BC132865.1 - 1 38 40146887
U65378.1 - 2 111
U89012.1 - 2 111

View Full Product Details