Gene Transcripts

Gene Symbol
IFNA5
Entrez Gene ID
3442
Gene Name
interferon alpha 5
Gene Aliases
IFN-alpha-5, IFN-alphaG, INA5, INFA5, leIF G
Chromosome Location
Chr. 9: 21304614 - 21305313 on Build GRCh38
UniGene ID
Hs.37113
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_002169.2 NP_002160.1 1 504
GenBank mRNA BC093755.1 - 1 504
BC093757.1 - 1 504 7939602
V00541.1 - 1 279

View Full Product Details