Gene Transcripts

Gene Symbol
INPP1
Entrez Gene ID
3628
Gene Name
inositol polyphosphate-1-phosphatase
Gene Aliases
Chromosome Location
Chr. 2: 190343470 - 190371665 on Build GRCh38
UniGene ID
Hs.32309
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001128928.1 NP_001122400.1 6 1210
NM_002194.3 NP_002185.1 5 1066
XM_005246532.1 XP_005246589.1 5 847
GenBank mRNA AB451298.1 - 4 510
AK093560.1 - 7 1168
BC015496.1 - 4 576 3845956
BC106006.1 - 5 610 4472604
CR456840.1 - 4 510
CR541707.1 - 4 510
KJ535023.1 - 4 510
KJ535042.1 - 4 509
KU178146.1 - 4 510
L08488.1 - 5 836

View Full Product Details