Gene Transcripts

Gene Symbol
PRKCG
Entrez Gene ID
5582
Gene Name
protein kinase C gamma
Gene Aliases
PKC-gamma, PKCC, PKCG, SCA14
Chromosome Location
Chr. 19: 53882184 - 53907647 on Build GRCh38
UniGene ID
Hs.631564
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001316329.1 NP_001303258.1 13 1705
NM_002739.4 NP_002730.1 13 1705
GenBank mRNA AF345987.1 - 13 1589
AK296412.1 - 13 1713
AK296434.1 - 13 1692
AK303741.1 - 11 1112
AK312302.1 - 13 1667
BC047876.1 - 13 1580 5764695
Z15114.1 - 9 913

View Full Product Details