Gene Transcripts

Gene Symbol
SGCA
Entrez Gene ID
6442
Gene Name
sarcoglycan alpha
Gene Aliases
50DAG, ADL, DAG2, DMDA2, LGMD2D, SCARMD1, adhalin
Chromosome Location
Chr. 17: 50165517 - 50175932 on Build GRCh38
UniGene ID
Hs.463412
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_000023.3 NP_000014.1 4 385
NM_001135697.2 NP_001129169.1 4 385
NR_135553.1 4 385
XM_011525120.2 XP_011523422.2 4 851
XM_011525121.2 XP_011523423.2 4 853
XM_011525122.2 XP_011523424.2 4 851
XM_011525123.2 XP_011523425.2 4 851
XM_011525124.2 XP_011523426.1 3 206
XM_017024950.1 XP_016880439.1 4 847
GenBank mRNA AK290622.1 - 4 358
AK293614.1 - 3 206
AK300710.1 - 4 367
BC025702.1 - 4 357 5222483
DC306167.1 - 3 289
KU178331.1 - 4 329
KU178332.1 - 4 329
L34355.1 - 4 366
L35853.1 - 4 366
L46810.1 - 4 366
U08895.1 - 4 340

View Full Product Details