Gene Transcripts

Gene Symbol
STC1
Entrez Gene ID
6781
Gene Name
stanniocalcin 1
Gene Aliases
STC
Chromosome Location
Chr. 8: 23841921 - 23854807 on Build GRCh38
UniGene ID
Hs.25590
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_003155.2 NP_003146.1 1 327
GenBank mRNA AK312589.1 - 1 326
BC029044.1 - 1 305 5191420
U25997.1 - 1 327
U46768.1 - 1 77

View Full Product Details