Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_020783.3 | NP_065834.1 | 2, 3 | 1216 | |
GenBank mRNA | AB037763.1 | - | 2, 3 | 1033 | |
AF299075.1 | - | 2, 3 | 940 | ||
AK291154.1 | - | 2, 3 | 1056 | ||
AK293791.1 | - | 2, 3 | 1002 | ||
AK297379.1 | - | 2, 3 | 293 | ||
AK299593.1 | - | 3, 4 | 459 | ||
AK299652.1 | - | 3, 4 | 712 | ||
BC036538.2 | - | 2, 3 | 945 | 5260337 |