Gene Transcripts

Gene Symbol
TGFBI
Entrez Gene ID
7045
Gene Name
transforming growth factor beta induced
Gene Aliases
BIGH3, CDB1, CDG2, CDGG1, CSD, CSD1, CSD2, CSD3, EBMD, LCD1
Chromosome Location
Chr. 5: 136028895 - 136063818 on Build GRCh38
UniGene ID
Hs.369397
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_000358.2 NP_000349.1 13 1894
GenBank mRNA AK093916.1 - 13 1749
AK094055.1 - 14 1825
AK094581.1 - 13 1838
AK222833.1 - 13 1802
AK298603.1 - 8 1114
AK303643.1 - 13 2126
BC000097.2 - 13 1787 3506051
BC004972.1 - 13 1782 2958878
BT009820.1 - 13 1733
KU178357.1 - 13 1733
KU178358.1 - 11 1247
M77349.1 - 13 1780

View Full Product Details