Gene Transcripts

Gene Symbol
TSPAN6
Entrez Gene ID
7105
Gene Name
tetraspanin 6
Gene Aliases
T245, TM4SF6, TSPAN-6
Chromosome Location
Chr. X: 100627108 - 100637104 on Build GRCh38
UniGene ID
Hs.43233
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001278740.1 NP_001265669.1 2 591
NM_001278741.1 NP_001265670.1 2 361
NM_001278742.1 NP_001265671.1 2 361
NM_001278743.1 NP_001265672.1 2 361
NM_003270.3 NP_003261.1 2 299
XM_011531018.1 XP_011529320.1 3 101
GenBank mRNA AB097016.1 - 2 195
AB209876.1 - 2 258
AF043906.1 - 2 203
AF053453.1 - 2 155
AF133426.1 - 2 162
AK075072.1 - 2 248
AK309581.1 - 2 574
AK315773.1 - 2 280
AY358825.1 - 2 176
BC012389.1 - 2 239 3857537
BT006977.1 - 2 136
CN274931.1 - 2 81
CR457115.1 - 2 136
CX760536.1 - 2 299
CX868833.1 - 2 70
DA038474.1 - 2 98
U84895.1 - 2 203

View Full Product Details