Gene Transcripts

Gene Symbol
TSNAX
Entrez Gene ID
7257
Gene Name
translin associated factor X
Gene Aliases
TRAX
Chromosome Location
Chr. 1: 231528653 - 231566524 on Build GRCh38
UniGene ID
Hs.13318
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_005999.2 NP_005990.1 2, 3 268
GenBank mRNA AK313068.1 - 2, 3 206
BC010376.1 - 2, 3 226 4283594
BC011797.1 - 2, 3 243 4138517
X95073.1 - 2, 3 270

Gene Symbol
TSNAX-DISC1
Entrez Gene ID
100303453
Gene Name
TSNAX-DISC1 readthrough (NMD candidate)
Gene Aliases
Chromosome Location
Chr. 1: 231528653 - 232041272 on Build GRCh38
UniGene ID
Hs.13318
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NR_028393.1 2, 3 268
NR_028394.1 2, 3 268
NR_028395.1 2, 3 268
NR_028396.1 2, 3 268
NR_028397.1 2, 3 268
NR_028399.1 2, 3 268
NR_028400.1 2, 3 268

View Full Product Details