Gene Transcripts
- Gene Symbol
- ZNF17
- Entrez Gene ID
- 7565
- Gene Name
- zinc finger protein 17
- Gene Aliases
- HPF3, KOX10
- Chromosome Location
- Chr. 19: 57409252 - 57421939 on Build GRCh38
- UniGene ID
- Hs.729403
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_006959.2 |
NP_008890.2
|
1, 2 |
244 |
|
GenBank mRNA |
AK098183.1 |
- |
1, 2 |
226 |
|
View Full Product Details