Gene Transcripts

Gene Symbol
ZNF70
Entrez Gene ID
7621
Gene Name
zinc finger protein 70
Gene Aliases
Cos17
Chromosome Location
Chr. 22: 23741584 - 23751092 on Build GRCh38
UniGene ID
Hs.382874
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_021916.3 NP_068735.1 2 666
GenBank mRNA AK122721.1 - 1 233
BC040161.1 - 2 666 5551072
CR456615.1 - 2 339

View Full Product Details