Gene Transcripts
- Gene Symbol
- REEP5
- Entrez Gene ID
- 7905
- Gene Name
- receptor accessory protein 5
- Gene Aliases
- C5orf18, D5S346, DP1, TB2, YOP1, Yip2e
- Chromosome Location
- Chr. 5: 112876384 - 112922334 on Build GRCh38
- UniGene ID
- Hs.429608
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_005669.4 |
NP_005660.4
|
4, 5 |
674 |
|
GenBank mRNA |
AK294496.1 |
- |
3, 4 |
660 |
|
AK295436.1 |
- |
3, 4 |
426 |
|
AK298268.1 |
- |
3, 4 |
409 |
|
AK299902.1 |
- |
5, 6 |
938 |
|
AK300048.1 |
- |
4, 5 |
492 |
|
AK310729.1 |
- |
4, 5 |
1038 |
|
AK312522.1 |
- |
4, 5 |
579 |
|
AY562243.1 |
- |
4, 5 |
530 |
|
BC000232.1 |
- |
4, 5 |
555 |
3350749 |
BC065926.1 |
- |
4, 5 |
533 |
6164621 |
BX648005.1 |
- |
4, 5 |
655 |
|
M74090.1 |
- |
4, 5 |
556 |
|
View Full Product Details