Gene Transcripts

Gene Symbol
ZNHIT3
Entrez Gene ID
9326
Gene Name
zinc finger HIT-type containing 3
Gene Aliases
TRIP3
Chromosome Location
Chr. 17: 36486627 - 36499310 on Build GRCh38
UniGene ID
Hs.2210
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001281432.1 NP_001268361.1 1 90
NM_001281433.1 NP_001268362.1 1 90
NM_001281434.1 NP_001268363.1 1 90
NM_004773.3 NP_004764.1 1 90
NR_104009.1 1 90
NR_104010.1 1 90
NR_104011.1 1 90
GenBank mRNA AF400652.1 - 1 82
AK290858.1 - 1 41
AK308583.1 - 1 18
AK310022.1 - 1 25
AV763423.1 - 57
BC017931.1 - 1 55 4669658
BC105608.1 - 1 62
BG032116.1 - 1 25
BI767446.1 - 1 23
BM457955.1 - 25
CB996852.1 - 1 115
HY045632.1 - 1 91

View Full Product Details