Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_004815.3 | NP_004806.3 | 23 | 3695 | |
XM_006711048.3 | XP_006711111.1 | 22 | 3329 | ||
XM_011542438.2 | XP_011540740.1 | 23 | 3570 | ||
XM_011542439.2 | XP_011540741.1 | 23 | 3415 | ||
XM_011542440.2 | XP_011540742.1 | 22 | 3289 | ||
GenBank mRNA | AB208873.1 | - | 22 | 3301 | |
BC093741.1 | - | 22 | 3368 | 7939586 | |
BC093767.1 | - | 22 | 3371 | 7939612 | |
CA454270.1 | - | 1 | 153 | ||
U90920.1 | - | 23 | 3509 |