Gene Transcripts

Gene Symbol
MLEC
Entrez Gene ID
9761
Gene Name
malectin
Gene Aliases
KIAA0152
Chromosome Location
Chr. 12: 120687124 - 120701864 on Build GRCh38
UniGene ID
Hs.744910
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001303627.1 NP_001290556.1 2 176
NM_001303628.1 NP_001290557.1 2 459
NM_014730.3 NP_055545.1 2 459
XM_011539031.1 XP_011537333.1 2 4874
XM_011539032.1 XP_011537334.1 3 4562
GenBank mRNA BC000371.2 - 2 408
BC016297.2 - 2 410 3832682
BE326824.1 - 2 160
BG718506.1 - 2 464
BI458950.1 - 2 178
BU845446.1 - 437
D63486.1 - 2 414

View Full Product Details