Gene Transcripts

Gene Symbol
RBX1
Entrez Gene ID
9978
Gene Name
ring-box 1
Gene Aliases
BA554C12.1, RNF75, ROC1
Chromosome Location
Chr. 22: 40951347 - 40973015 on Build GRCh38
UniGene ID
Hs.474949
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_014248.3 NP_055063.1 2, 3 200
GenBank mRNA AF085906.1 - 2, 3 129
AF140598.1 - 2, 3 154
AF142059.1 - 2, 3 148
AK315722.1 - 2, 3 159
AY099360.1 - 2, 3 111
BC001466.2 - 2, 3 153 3138751
BC017370.1 - 2, 3 170
CR456560.1 - 2, 3 166
DA353856.1 - 2, 3 200

View Full Product Details