Gene Transcripts
- Gene Symbol
- TOM1
- Entrez Gene ID
- 10043
- Gene Name
- target of myb1 membrane trafficking protein
- Gene Aliases
- Chromosome Location
- Chr. 22: 35299275 - 35347994 on Build GRCh38
- UniGene ID
- Hs.474705
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001135729.1 |
NP_001129201.1
|
13, 14 |
1344 |
|
NM_001135730.1 |
NP_001129202.1
|
12, 13 |
1282 |
|
NM_001135732.1 |
NP_001129204.1
|
13, 14 |
1417 |
|
NM_005488.2 |
NP_005479.1
|
13, 14 |
1417 |
|
NR_024194.1 |
|
13, 14 |
1446 |
|
NR_024195.1 |
|
12, 13 |
1367 |
|
XM_011529818.2 |
XP_011528120.1
|
14, 15 |
1431 |
|
XM_011529820.2 |
XP_011528122.1
|
12, 13 |
1282 |
|
XM_017028529.1 |
XP_016884018.1
|
14, 15 |
1431 |
|
XM_017028530.1 |
XP_016884019.1
|
12, 13 |
1282 |
|
GenBank mRNA |
AJ006973.1 |
- |
13, 14 |
1353 |
|
AK090723.1 |
- |
1, 2 |
926 |
|
AK097066.1 |
- |
11, 12 |
928 |
|
AK097926.1 |
- |
15, 16 |
1633 |
|
AK293694.1 |
- |
13, 14 |
1344 |
|
AK295918.1 |
- |
12, 13 |
1277 |
|
AK296670.1 |
- |
13, 14 |
1354 |
|
AK297827.1 |
- |
12, 13 |
1196 |
|
AK314522.1 |
- |
13, 14 |
1326 |
|
BC040006.1 |
- |
13, 14 |
1310 |
|
BC046151.1 |
- |
13, 14 |
1295 |
6470390 |
View Full Product Details