Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001300823.1 | NP_001287752.1 | 12 | 1671 | |
NM_001300824.1 | NP_001287753.1 | 12 | 1671 | ||
NM_018699.3 | NP_061169.2 | 13 | 1764 | ||
XM_011531562.2 | XP_011529864.1 | 13 | 1780 | ||
XM_011531563.2 | XP_011529865.1 | 12 | 1687 | ||
XM_011531564.2 | XP_011529866.1 | 13 | 1780 | ||
XM_011531565.2 | XP_011529867.1 | 13 | 1780 | ||
XM_011531566.2 | XP_011529868.1 | 13 | 1780 | ||
XM_011531567.2 | XP_011529869.1 | 13 | 1780 | ||
XM_011531568.2 | XP_011529870.1 | 13 | 1780 | ||
XM_011531569.2 | XP_011529871.1 | 13 | 1780 | ||
XM_011531570.2 | XP_011529872.1 | 11 | 1397 | ||
XM_017007668.1 | XP_016863157.1 | 13 | 2273 | ||
XM_017007669.1 | XP_016863158.1 | 13 | 1747 | ||
XM_017007670.1 | XP_016863159.1 | 13 | 1747 | ||
XM_017007671.1 | XP_016863160.1 | 7 | 871 | ||
GenBank mRNA | AF272897.1 | - | 13 | 1602 | |
AK314293.1 | - | 12 | 1837 | ||
BC121037.1 | - | 12 | 1458 | 40117856 | |
BC121038.1 | - | 12 | 1458 | 40117857 |