Gene Transcripts

Gene Symbol
OR1A2
Entrez Gene ID
26189
Gene Name
olfactory receptor family 1 subfamily A member 2
Gene Aliases
OR17-6
Chromosome Location
Chr. 17: 3197517 - 3198448 on Build GRCh38
UniGene ID
Hs.532660
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_012352.2 NP_036484.1 1 422
GenBank mRNA BC069092.1 - 1 422
BC106733.1 - 1 420 40015715
BC106734.1 - 1 420 40015716

View Full Product Details