Gene Transcripts
- Gene Symbol
- ARFIP1
- Entrez Gene ID
- 27236
- Gene Name
- ADP ribosylation factor interacting protein 1
- Gene Aliases
- HSU52521
- Chromosome Location
- Chr. 4: 152779937 - 152911912 on Build GRCh38
- UniGene ID
- Hs.416089
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001025593.2 |
NP_001020764.1
|
4, 5 |
617 |
|
NM_001025595.2 |
NP_001020766.1
|
5, 6 |
713 |
|
NM_001287431.1 |
NP_001274360.1
|
5, 6 |
578 |
|
NM_001287432.1 |
NP_001274361.1
|
6, 7 |
830 |
|
NM_001287433.1 |
NP_001274362.1
|
5, 6 |
599 |
|
NM_014447.3 |
NP_055262.1
|
4, 5 |
482 |
|
XM_011531868.2 |
XP_011530170.1
|
4, 5 |
3540 |
|
XM_011531869.2 |
XP_011530171.1
|
6, 7 |
727 |
|
XM_011531870.2 |
XP_011530172.1
|
6, 7 |
592 |
|
XM_011531871.2 |
XP_011530173.1
|
3, 4 |
3446 |
|
XM_011531872.1 |
XP_011530174.1
|
4, 5 |
385 |
|
GenBank mRNA |
AF124489.1 |
- |
4, 5 |
414 |
|
AK290472.1 |
- |
4, 5 |
453 |
|
AK304141.1 |
- |
5, 6 |
578 |
|
AK316125.1 |
- |
3, 4 |
327 |
|
AU139844.1 |
- |
6, 7 |
572 |
|
BC103759.1 |
- |
5, 6 |
542 |
6176474 |
BC105114.1 |
- |
4, 5 |
569 |
8144117 |
BC105116.1 |
- |
4, 5 |
569 |
8144119 |
BC143675.1 |
- |
4, 5 |
569 |
9052189 |
BC143676.1 |
- |
5, 6 |
530 |
9052190 |
CB127228.1 |
- |
5, 6 |
505 |
|
CN305403.1 |
- |
4, 5 |
565 |
|
U52521.1 |
- |
4, 5 |
448 |
|
View Full Product Details