Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001199492.1 | NP_001186421.1 | 3 | 397 | |
NM_014456.4 | NP_055271.2 | 3 | 439 | ||
NM_145341.3 | NP_663314.1 | 4 | 523 | ||
GenBank mRNA | AK223623.1 | - | 3 | 313 | |
AK296753.1 | - | 3 | 264 | ||
AK312540.1 | - | 3 | 335 | ||
AK315295.1 | - | 4 | 401 | ||
BC015036.2 | - | 3 | 303 | ||
BI910668.1 | - | 3 | 401 | ||
BX537500.1 | - | 2 | 3387 | ||
BX647834.1 | - | 3 | 283 | ||
DB241608.1 | - | 3 | 440 | ||
U96628.1 | - | 3 | 237 |