Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_013313.4 | NP_037445.1 | 5 | 718 | |
NR_130910.1 | 5 | 1102 | |||
GenBank mRNA | AB098734.1 | - | 5 | 718 | |
AB098735.1 | - | 5 | 718 | ||
AF060862.1 | - | 4 | 418 | ||
AF305069.1 | - | 4 | 478 | ||
AW006162.1 | - | 1 | 290 | ||
BC034486.1 | - | 5 | 576 | 4826997 | |
BI559478.1 | - | 5 | 597 | ||
CR456376.2 | - | 4 | 365 | ||
CR933630.1 | - | 4 | 4821 |