Gene Transcripts
- Gene Symbol
- SIDT2
- Entrez Gene ID
- 51092
- Gene Name
- SID1 transmembrane family member 2
- Gene Aliases
- CGI-40
- Chromosome Location
- Chr. 11: 117179223 - 117197445 on Build GRCh38
- UniGene ID
- Hs.712144
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001040455.1 |
NP_001035545.1
|
16, 17 |
1534 |
|
GenBank mRNA |
AF151799.1 |
- |
17, 18 |
1639 |
|
AK075137.1 |
- |
7, 8 |
512 |
|
AK292501.1 |
- |
16, 17 |
2014 |
|
AK292974.1 |
- |
16, 17 |
1525 |
|
AK303023.1 |
- |
16, 17 |
1490 |
|
AY358442.1 |
- |
16, 17 |
1543 |
|
BC114522.1 |
- |
17, 18 |
1554 |
40035836 |
BX648437.1 |
- |
8, 9 |
632 |
|
KC109742.1 |
- |
16, 17 |
1490 |
|
View Full Product Details