Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_016234.3 | NP_057318.2 | 13 | 1453 | |
NM_203379.1 | NP_976313.1 | 13 | 1314 | ||
NM_203380.1 | NP_976314.1 | 13 | 1480 | ||
XM_017016324.1 | XP_016871813.1 | 13 | 1488 | ||
GenBank mRNA | AB033899.1 | - | 13 | 1327 | |
AK000339.1 | - | 13 | 1463 | ||
AK024932.1 | - | 13 | 1454 | ||
AK222782.1 | - | 13 | 1471 | ||
AK223288.1 | - | 13 | 1442 | ||
AK301786.1 | - | 8 | 683 | ||
AK310436.1 | - | 13 | 1263 | ||
AM262166.1 | - | 12 | 1173 | ||
AY358520.1 | - | 13 | 1414 | ||
BC007985.2 | - | 13 | 1431 | 2959548 |