Gene Transcripts

Gene Symbol
GIN1
Entrez Gene ID
54826
Gene Name
gypsy retrotransposon integrase 1
Gene Aliases
GIN-1, TGIN1, ZH2C2
Chromosome Location
Chr. 5: 103086000 - 103120149 on Build GRCh38
UniGene ID
Hs.24088
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001317954.1 NP_001304883.1 5 801
NM_017676.2 NP_060146.2 6 995
XM_005272028.3 XP_005272085.1 6 1002
XM_017009599.1 XP_016865088.1 6 2430
GenBank mRNA AK000132.1 - 6 993
AK225572.1 - 6 992
AK295796.1 - 6 983
BC151135.1 - 5 935 9057035
BC157835.1 - 5 935 9057054
CR627424.1 - 5 772

View Full Product Details