Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_017741.3 | NP_060211.3 | 3 | 1161 | |
XM_005248169.3 | XP_005248226.1 | 3 | 1149 | ||
XM_005248171.3 | XP_005248228.1 | 2 | 1045 | ||
XM_006713966.3 | XP_006714029.1 | 4 | 1320 | ||
XM_017008327.1 | XP_016863816.1 | 2 | 2614 | ||
XM_017008328.1 | XP_016863817.1 | 2 | 2599 | ||
GenBank mRNA | AK000287.1 | - | 1 | 283 | |
AK056116.1 | - | 3 | 1161 | ||
BC050697.1 | - | 2 | 822 | 6044417 | |
BC068025.1 | - | 1 | 460 | 4593954 | |
BC101716.1 | - | 1 | 171 | 8069222 | |
BC101718.1 | - | 1 | 171 | 8069224 |