Gene Transcripts
- Gene Symbol
- WDR70
- Entrez Gene ID
- 55100
- Gene Name
- WD repeat domain 70
- Gene Aliases
- Chromosome Location
- Chr. 5: 37379280 - 37752672 on Build GRCh38
- UniGene ID
- Hs.213690
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_018034.2 |
NP_060504.1
|
10, 11 |
1158 |
|
XM_005248318.3 |
XP_005248375.1
|
10, 11 |
1176 |
|
XM_011514061.2 |
XP_011512363.1
|
4, 5 |
471 |
|
XM_017009604.1 |
XP_016865093.1
|
9, 10 |
992 |
|
XM_017009605.1 |
XP_016865094.1
|
9, 10 |
959 |
|
GenBank mRNA |
AK001095.1 |
- |
10, 11 |
1100 |
|
AK291179.1 |
- |
10, 11 |
1180 |
|
AL512685.1 |
- |
10, 11 |
1159 |
|
BC009648.1 |
- |
10, 11 |
1158 |
3454326 |
BC025315.1 |
- |
10, 11 |
1136 |
5458850 |
BX537655.1 |
- |
9, 10 |
1115 |
|
View Full Product Details