Gene Transcripts

Gene Symbol
VPS13D
Entrez Gene ID
55187
Gene Name
vacuolar protein sorting 13 homolog D
Gene Aliases
Chromosome Location
Chr. 1: 12230039 - 12512047 on Build GRCh38
UniGene ID
Hs.439381
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_015378.3 NP_056193.2 70 13265
NM_018156.3 NP_060626.2 69 13190
GenBank mRNA AB007922.3 - 52 9572
AJ608774.1 - 70 13237
AJ608775.1 - 69 13162
AK026719.1 - 4 758
AK074350.1 - 11 1583
AK095438.1 - 1 672
AK295882.1 - 8 1147
AK304850.1 - 4 770
AL833079.1 - 25 3876
AL833320.1 - 3591
BC028115.1 - 28 4181
BC051804.1 - 18 2667
BX641035.1 - 44 6753

View Full Product Details