Gene Transcripts
- Gene Symbol
- MCOLN3
- Entrez Gene ID
- 55283
- Gene Name
- mucolipin 3
- Gene Aliases
- TRP-ML3, TRPML3
- Chromosome Location
- Chr. 1: 85018082 - 85048902 on Build GRCh38
- UniGene ID
- Hs.535239
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001253693.1 |
NP_001240622.1
|
8, 9 |
1075 |
|
NM_018298.10 |
NP_060768.8
|
9, 10 |
1243 |
|
XM_005271003.1 |
XP_005271060.1
|
10, 11 |
1640 |
|
XM_006710750.1 |
XP_006710813.1
|
10, 11 |
1459 |
|
XM_011541739.2 |
XP_011540041.2
|
9, 10 |
1167 |
|
XM_011541740.2 |
XP_011540042.1
|
8, 9 |
1001 |
|
XM_011541742.1 |
XP_011540044.1
|
10, 11 |
1377 |
|
GenBank mRNA |
AF475085.1 |
- |
9, 10 |
1149 |
|
AK001868.1 |
- |
8, 9 |
981 |
|
AK292206.1 |
- |
9, 10 |
1175 |
|
AK314354.1 |
- |
9, 10 |
1203 |
|
View Full Product Details